Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

900189

Sigma-Aldrich

Poly(N-isopropyl acylamide-co-3-(trimethoxysilyl)propyl methacrylate)

5-10% (trimethoxysilyl)propyl methacrylate, average Mn 10000

Synonym(s):

PNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

powder

mol wt

average Mn 10000

impurities

5-10% (trimethoxysilyl)propyl methacrylate

Mw/Mn

<3

storage temp.

−20°C

Application

This polymer is thermosensitive and can be coated on glass, silica, iron oxide, or other oxide nanoparticles, larger particles, or other surfaces. PNIPAM based polymers are typically used in biomedical applications such as in cell sheet culture and tissue engineering.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service