Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900187

Sigma-Aldrich

Poly(9,9-dioctyl-9H-fluorene-2,7-diyl)

3,5-dimethylphenyl terminated

Synonym(s):

Poly(9,9-di-n-octylfluorenyl-2,7-diyl), PFO, Poly(9,9- dioctylfluorene), Poly(9,9-di-n-octyl-2,7-fluorene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H9(C29H40)nC8H9
MDL number:
UNSPSC Code:
12352200

form

powder (or fibres)

mol wt

average Mn >10,000

PDI

<4.0

Looking for similar products? Visit Product Comparison Guide

Application

Polyfluorenes (PFs) are one of the most important PLED materials, showing a strong blue fluorescence stabilized by its rigid, conjugated structure.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malti Bansal et al.
Nanoscale, 2(7), 1171-1177 (2010-07-22)
Multiwall carbon nanotubes (MWNTs) have been added to the polymer poly (9,9-dioctylfluorenyl-2, 7-diyl) end capped with dimethylphenyl (PFO) in various weight percentages and the blends thus prepared, using a solution processing approach, have been characterized using SEM, UV-VIS spectroscopy, PL
Large enhancement of intersystem crossing in polyfluorenes by iridium-complex doping.
Liao et al.
Applied Physics Letters, 90(1), 013504/1-013504/3 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service