Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900186

Sigma-Aldrich

1,7-Dibromo-3,4,9,10-tetracarboxylic acid dianhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H6Br2O6
CAS Number:
Molecular Weight:
550.11
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

assay

98%

form

powder

SMILES string

O=C(C(C=C1)=C2C3=C1C4=C5C(C3=C(Br)C=C26)=CC=C7C5=C(C(OC7=O)=O)C=C4Br)OC6=O

InChI

1S/C24H6Br2O6/c25-13-5-12-16-10(22(28)32-24(12)30)4-2-8-18-14(26)6-11-15-9(21(27)31-23(11)29)3-1-7(19(15)18)17(13)20(8)16/h1-6H

InChI key

WPBVUAVIGWNDGT-UHFFFAOYSA-N

Application

Synthetic building block of high-mobility air-stable n-type semiconductors (electron-transport small organic molecules and polymers).

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved Performance of Solution-Processed n-Type Organic Field-Effect Transistors by Regulating the Intermolecular Interactions and Crystalline Domains on Macroscopic Scale.
Vasimalla et al.
Chemistry of Materials, 26(13), 4030-4037 (2014)
A high-mobility electron-transport polymer with broad absorption and its use in field-effect transistors and all-polymer solar cells.
Xiaowei Zhan et al.
Journal of the American Chemical Society, 129(23), 7246-7247 (2007-05-19)
A water/alcohol-soluble copolymer based on fluorene and perylene diimide as a cathode interlayer for inverted polymer solar cells.
Zhao et al.
Journal of Material Chemistry C, 3(17), 4515-4521 (2015)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service