900147
Iron oxide (II,III), nanoparticles
5 nm avg. part. size (TEM), dextran functionalized, 10 mg/mL in H2O
About This Item
Recommended Products
form
dispersion
nanoparticles
concentration
10 mg/mL in H2O
avg. part. size
5 nm (TEM)
density
1 g/cm3
functional group
dextran
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.
Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.
Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service