900146
Iron oxide (II,III), nanoparticles
10 nm avg. part. size (TEM), rhodamine B functionalized, 1 mg/mL in H2O
About This Item
Recommended Products
form
dispersion
nanoparticles
concentration
1 mg/mL in H2O
avg. part. size
10 nm (TEM)
functional group
rhodamine B
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Nanoparticle imaging agents enable stem cell tracking via various in vivo imaging techniques.
Professor Hui Mao explores the use of superparamagnetic iron oxide nanoparticles (INOPs) that offer an alternate contrast-enhancing mechanism.
Prof. Yadong Yin discusses various synthesis methods of magnetite nanocrystals and their applications in different fields.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service