Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89753

Sigma-Aldrich

Phenyldi(o-tolyl)phosphine polymer-bound

extent of labeling: ~2.2 mmol/g loading

Synonym(s):

Bis(2-methylphenyl)phenylphosphine polymer-bound, Di(o-tolyl)phosphino-polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: ligand

extent of labeling

~2.2 mmol/g loading

particle size

100-200 mesh

functional group

phosphine

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service