Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87462

Sigma-Aldrich

(1S,2R)-1-Methyl cis-1,2,3,6-tetrahydrophthalate

≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

(1S,2R)-1-Methyl cis-4-cyclohexene-1,2-dicarboxylate, (1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CC6H8CO2CH3
CAS Number:
Molecular Weight:
184.19
Beilstein/REAXYS Number:
5262096
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (sum of enantiomers, HPLC)

form

crystals

optical activity

[α]20/D +11.5±1°, c = 1% in acetone

mp

65-67 °C (lit.)

SMILES string

COC(=O)[C@H]1CC=CC[C@H]1C(O)=O

InChI

1S/C9H12O4/c1-13-9(12)7-5-3-2-4-6(7)8(10)11/h2-3,6-7H,4-5H2,1H3,(H,10,11)/t6-,7+/m1/s1

InChI key

MYYLMIDEMAPSGH-RQJHMYQMSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral building block used for the synthesis of chiral cyclohexane compounds, e.g. carbapenems[1]; Pheromones[2]; etc.[3][4]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.S. Hamilton et al.
The Journal of Organic Chemistry, 58, 7263-7263 (1993)
N. Tamura et al.
Tetrahedron Letters, 27, 3749-3749 (1986)
W. Boland et al.
Helvetica Chimica Acta, 68, 2062-2062 (1985)
S. Kobayashi et al.
Tetrahedron Letters, 25, 2557-2557 (1984)
S Kobayashi et al.
Chemical & pharmaceutical bulletin, 38(2), 350-354 (1990-02-01)
A versatile chiral synthon, (1R,6S)-6-methoxycarbonyl-3-cyclohexene-1-carboxylic acid, was obtained by an enantioselective hydrolysis of the corresponding meso diester with pig liver esterase. This enzymatic hydrolysis can easily be carried out on a multi-hundred gram scale. The chiral monoester thus obtained can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service