86487
(±)-α-Terpinyl acetate, predominantly α-isomer
technical, ≥90% (GC)
Synonym(s):
(±)-2-(4-Methyl-3-cyclohexenyl)isopropyl acetate
About This Item
Recommended Products
grade
technical
assay
≥90% (GC)
refractive index
n20/D 1.465 (lit.)
bp
220 °C (lit.)
density
0.953 g/mL at 25 °C (lit.)
SMILES string
CC(=O)OC(C)(C)C1CCC(C)=CC1
InChI
1S/C12H20O2/c1-9-5-7-11(8-6-9)12(3,4)14-10(2)13/h5,11H,6-8H2,1-4H3
InChI key
IGODOXYLBBXFDW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service