The trityl group can be removed with 95 percent TFA (trifluoroacetic acid). This will generate a free sulfhydryl (-SH) group. We do not have details on the reaction (time, temperature amount of TFA to use) from the original manufacturer.
86343
TentaGel™ HL-S-Trityl
extent of labeling: ~0.40 mmol/g loading
Synonym(s):
O-[2-(3-Tritylthio-propionamido)ethyl]polyethylene glycol polymer-bound
About This Item
Recommended Products
product line
TentaGel™
form
solid
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
extent of labeling
~0.40 mmol/g loading
particle size
~110 μm
storage temp.
2-8°C
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can the trityl group be removed from 86343 TentaGel™ HL-S-Trityl?
1 answer-
Helpful?
-
-
For Product 86343 TentaGel™ HL-S-Trityl, what is the best way to deprotect the sulfur (that is, to remove the trityl group)?
1 answer-
TentaGel is a trademark of Rapp Polymere GmbH (in Germany). They have provided the following information:One can remove the trityl group with 95% TFA.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service