Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860492

Sigma-Aldrich

2-Naphthyl-β-D-galactopyranoside hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O6 · xH2O
CAS Number:
Molecular Weight:
306.31 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

crystals

optical activity

[α]20/D −55°, c = 1 in ethanol

storage temp.

−20°C

SMILES string

[H]O[H].OC[C@H]1OC(Oc2ccc3ccccc3c2)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C16H18O6.H2O/c17-8-12-13(18)14(19)15(20)16(22-12)21-11-6-5-9-3-1-2-4-10(9)7-11;/h1-7,12-20H,8H2;1H2/t12-,13+,14+,15-,16-;/m1./s1

InChI key

GFPMJKVBHWMQGT-WKILGUOFSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Liang et al.
Cancer cell, 23(1), 107-120 (2013-01-01)
Inflammatory bowel disease is an important risk factor for colorectal cancer. We show that sphingosine-1-phosphate (S1P) produced by upregulation of sphingosine kinase 1 (SphK1) links chronic intestinal inflammation to colitis-associated cancer (CAC) and both are exacerbated by deletion of Sphk2.
Ratna Karuna et al.
Atherosclerosis, 219(2), 855-863 (2011-09-29)
Apolipoprotein M (apoM) has been identified as a specific sphingosine-1-phosphate (S1P) binding protein of HDL. To investigate the in vivo effects of disturbed apoM or HDL metabolism we quantified S1P and apoM in plasmas of wild-type, apoM-knock-out, and apoM transgenic
Hazem J Abuhusain et al.
The Journal of biological chemistry, 288(52), 37355-37364 (2013-11-23)
Studies in cell culture and mouse models of cancer have indicated that the soluble sphingolipid metabolite sphingosine 1-phosphate (S1P) promotes cancer cell proliferation, survival, invasiveness, and tumor angiogenesis. In contrast, its metabolic precursor ceramide is prodifferentiative and proapoptotic. To determine
Maria L Allende et al.
The Journal of biological chemistry, 288(25), 18381-18391 (2013-05-03)
Sphingosine 1-phosphate (S1P) is a bioactive lipid whose levels are tightly regulated by its synthesis and degradation. Intracellularly, S1P is dephosphorylated by the actions of two S1P-specific phosphatases, sphingosine-1-phosphate phosphatases 1 and 2. To identify the physiological functions of S1P

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service