Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860441

Sigma-Aldrich

Nitrofurantoin

97%

Synonym(s):

N-(5-Nitro-2-furfurylidene)-1-aminohydantoin, Furadoxyl, Nitrofurantoine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N4O5
CAS Number:
Molecular Weight:
238.16
Beilstein/REAXYS Number:
893207
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

assay

97%

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(\C=N\N2CC(=O)NC2=O)o1

InChI

1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+

InChI key

NXFQHRVNIOXGAQ-YCRREMRBSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giacomo Russo et al.
International journal of pharmaceutics, 560, 294-305 (2019-02-17)
The present work proposes a novel application of EPSA (not an acronym but found to be referred to by many as Exposed Polar Surface Area), a supercritical fluid chromatography (SFC) polarity readout for assisting in the prediction of the extent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service