Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

857556

Sigma-Aldrich

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

mp

94-96 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CCO\C=C1/N=C(OC1=O)c2ccccc2

InChI

1S/C12H11NO3/c1-2-15-8-10-12(14)16-11(13-10)9-6-4-3-5-7-9/h3-8H,2H2,1H3/b10-8-

InChI key

SJHPCNCNNSSLPL-NTMALXAHSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Pradeille et al.
Chemistry & biodiversity, 9(11), 2528-2558 (2012-11-20)
The total syntheses of hypomurocin A3 and hypomuricin A5 (HM A3 and HM A5, resp.) in solution phase are described. These syntheses have been successfully achieved by applying the 'azirine/oxazolone method' to introduce the two Aib-Pro units into the backbone
Sylvère Durand et al.
The journal of physical chemistry. A, 117(12), 2508-2516 (2013-02-27)
The structure of peptide fragments was studied using "action" IR spectroscopy. We report on room temperature IR spectra of b4 fragments of protonated GGGGG, AAAAA, and YGGFL in the X-H (X = C, N, O) stretching region. Experiments were performed
Torsten Olszak et al.
Nature, 509(7501), 497-502 (2014-04-11)
The mechanisms by which mucosal homeostasis is maintained are of central importance to inflammatory bowel disease. Critical to these processes is the intestinal epithelial cell (IEC), which regulates immune responses at the interface between the commensal microbiota and the host.
Mikhail Kalinichev et al.
The Journal of pharmacology and experimental therapeutics, 344(3), 624-636 (2012-12-22)
Metabotropic glutamate receptor 7 (mGlu(7)) has been suggested to be a promising novel target for treatment of a range of disorders, including anxiety, post-traumatic stress disorder, depression, drug abuse, and schizophrenia. Here we characterized a potent and selective mGlu(7) negative
Michael J Rosen et al.
Journal of immunology (Baltimore, Md. : 1950), 190(4), 1849-1858 (2013-01-11)
Patients suffering from ulcerative colitis (UC) exhibit chronic colonic inflammation caused by a dysregulated mucosal immune response and epithelial barrier disruption. Th2 cytokines, including IL-13, have been implicated in the pathogenesis of UC. IL-13 induces phosphorylation of STAT6, and we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service