Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85400

Sigma-Aldrich

Silicone grease Bayer

low vacuum

Synonym(s):

Baysilone®

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100

packaging

tube of 35 g

Looking for similar products? Visit Product Comparison Guide

Legal Information

Baysilone is a registered trademark of Bayer AG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service