Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

850942

Sigma-Aldrich

DL-Ala-DL-Ala

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O3
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

SMILES string

CC(N)C(=O)NC(C)C(O)=O

InChI

1S/C6H12N2O3/c1-3(7)5(9)8-4(2)6(10)11/h3-4H,7H2,1-2H3,(H,8,9)(H,10,11)

InChI key

DEFJQIDDEAULHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes G Beck et al.
Journal of the American Chemical Society, 134(29), 12125-12133 (2012-06-29)
Insufficient oral bioavailability is considered as a key limitation for the widespread development of peptides as therapeutics. While the oral bioavailability of small organic compounds is often estimated from simple rules, similar rules do not apply to peptides, and even
Jessem Landoulsi et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(7), 1310-1316 (2011-04-14)
We investigate the interaction between D-Ala-D-Ala peptide and a stainless steel (SS) surface by AFM force spectroscopy with view to understand the role and nature of interfacial processes at the single molecule level. For this purpose, force-distance curves were recorded
Xiongwu Wu et al.
The Journal of chemical physics, 135(20), 204101-204101 (2011-12-02)
The self-guided Langevin dynamics (SGLD) is a method to accelerate conformational searching. This method is unique in the way that it selectively enhances and suppresses molecular motions based on their frequency to accelerate conformational searching without modifying energy surfaces or
Raphaël Plasson et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 41(5), 413-435 (2011-05-13)
A good comprehension of the reactivity of peptides in aqueous solution is fundamental in prebiotic chemistry, namely for understanding their stability and behavior in primitive oceans. Relying on the stereoselectivity of the involved reactions, there is a huge interest in
Andrew L Ferguson et al.
The Journal of chemical physics, 134(13), 135103-135103 (2011-04-12)
Nonlinear dimensionality reduction techniques can be applied to molecular simulation trajectories to systematically extract a small number of variables with which to parametrize the important dynamical motions of the system. For molecular systems exhibiting free energy barriers exceeding a few

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service