Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84222

Sigma-Aldrich

(R)-Fmoc-Nip-OH

≥99.0% (HPLC)

Synonym(s):

(R)-N-Fmoc-piperidine-3-carboxylic acid, (R)-Fmoc-nipecotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
Beilstein/REAXYS Number:
7945846
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1CCCN(C1)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C21H21NO4/c23-20(24)14-6-5-11-22(12-14)21(25)26-13-19-17-9-3-1-7-15(17)16-8-2-4-10-18(16)19/h1-4,7-10,14,19H,5-6,11-13H2,(H,23,24)/t14-/m1/s1

InChI key

FINXGQXNIBNREL-CQSZACIVSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service