84094
Methylthiourea bound to polystyrene
extent of labeling: ~2.0 mmol/g loading
Synonym(s):
N-Benzyl-thiourea polymer-bound, Thiourea polymer-bound
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
form
crystals
extent of labeling
~2.0 mmol/g loading
matrix
crosslinked with 2% DVB
particle size
200-400 mesh
storage temp.
2-8°C
Other Notes
Scavenger for active alkyl halides
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J S Warmus et al.
Bioorganic & medicinal chemistry letters, 8(17), 2309-2314 (1999-01-05)
Optimization of a 2-step reaction sequence was accomplished in 3-4 days, with over 200 different reaction conditions evaluated. Combinatorial arrays were performed using the optimized conditions to synthesize 590 new compounds which were tested for inhibition against N-His (D381E) ICE.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service