Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83558

Sigma-Aldrich

REM resin

extent of labeling: ~0.9 mmol/g loading

Synonym(s):

4-(Benzyloxy)benzyl acrylate, polymer-bound, Acrylate resin, Acryloyl Wang resin, Benzyl acrylate polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive

extent of labeling

~0.9 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

100-300 mesh

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

This resin allows the traceless synthesis of tertiary amines from secondary (or primary) amines. The resin can be regenerated; amine is coupled to the resin by Michael addition

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.R. Brown et al.
Journal of the American Chemical Society, 119, 3288-3288 (1997)
J.R. Morphy et al.
Tetrahedron Letters, 37, 3209-3209 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service