82904
(±)-α-Methyl-4-pyridinemethanol
≥98.0% (GC)
Synonym(s):
(±)-1-(4-Pyridyl)ethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
Beilstein/REAXYS Number:
109820
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
assay
≥98.0% (GC)
form
powder
mp
51-55 °C
functional group
hydroxyl
storage temp.
2-8°C
SMILES string
CC(O)c1ccncc1
InChI
1S/C7H9NO/c1-6(9)7-2-4-8-5-3-7/h2-6,9H,1H3
InChI key
HVOAMIOKNARIMR-UHFFFAOYSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service