Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82805

Sigma-Aldrich

Pyridinium chlorochromate polymer-bound

extent of labeling: ~3.5 mmol/g ClCrO3 loading

Synonym(s):

PVPCC, Poly(4-vinylpyridinium chlorochromate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

extent of labeling

~3.5 mmol/g ClCrO3 loading

matrix

crosslinked with 2% DVB

particle size

100-200 mesh

Other Notes

Polymeric recyclable oxidizing agent[1][2]

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.M.J. Frechet et al
The Journal of Organic Chemistry, 43, 2618-2618 (1978)
D.E. Bergbreiter et al.
Journal of the American Chemical Society, 107, 4792-4792 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service