Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

82795

Sigma-Aldrich

Pyridinium tribromide polymer-bound

extent of labeling: ~2 mmol/g Br3 loading

Synonym(s):

PVPHP, Poly(4-vinylpyridine hydrobromide perbromide), Pyridine hydrobromide perbromide polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

lumps

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~2 mmol/g Br3 loading

loss

≤2% loss on drying

matrix

crosslinked with 2% DVB

particle size

~60 mesh

SMILES string

Br[Br-]Br.c1cc[nH+]cc1

Other Notes

Easy to handle, stable and regenerable polymeric brominating agent, useful for the bromination of alkenes and ketones[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.M.J. Frechet et al.
J. Macromol. Sci. Chem., A11, 507-507 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service