Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81538

Sigma-Aldrich

Polystyrene A-CH(OEt)2

extent of labeling: 0.8-1.2 mmol/g loading

Synonym(s):

[2-(6,6-Diethoxyhexanoylamino)ethyl]polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive

extent of labeling

0.8-1.2 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

500-560 μm

functional group

aldehyde

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service