Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809624

Sigma-Aldrich

Gas Generator Kit

Synonym(s):

SyTracks GasBox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service