Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809411

Sigma-Aldrich

Lithium hexafluorophosphate solution

in dimethyl carbonate, 2.0 M LiPF6 DMC, battery grade

Synonym(s):

2 M LiPF6 DMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

form

solution

concentration

(2.0 M LiPF6 in DMC)

impurities

≤15 ppm H2O
≤50 ppm HF

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤2 ppm

cation traces

Ca: ≤1 ppm
Fe: ≤1 ppm
K: ≤1 ppm
Na: ≤1 ppm
Pb: ≤1 ppm

application(s)

battery manufacturing

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hexafluorophosphate solution in dimethyl carbonate is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.[1][2][3][4]

Application

The solutions of LiPF6 in alkyl carbonates find applications as electrolytes in lithium ion batteries[5][6] and the use of high quality battery grade electrolytes having extremely low water (<15 ppm) and hydrogen fluoride (<50 ppm) contents are critical for achieving high electrochemical performance.

Legal Information

Product of MU Ionic Solutions Corp

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation

target_organs

Bone,Teeth

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

68.0 °F

flash_point_c

20 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu G, et al.
Journal of Material Chemistry A, 3, 4092-4092 (2015)
Lamb J, et al.
Journal of the Electrochemical Society, 162, A2131-A2131 (2015)
The Li-ion rechargeable battery: a perspective
Goodenough JB and Park K
Journal of the American Chemical Society, 135(4), 1167-1176 (2013)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Articles

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Due to the adverse impact of the continued use of fossil fuels on the earth’s environment and climate, researchers have been asked to develop new approaches for producing power using renewable sources like wind and solar energy

Ionic liquid electrolytes explored for rechargeable batteries' advancement; future IL development discussed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service