Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809276

Sigma-Aldrich

Tetrahydrocortisone-2,2,3,4,4-d5 solution

100 μg/mL in methanol, ≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$477.85

$477.85

List Price$503.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request more information

Select a Size

Change View
1 ML
$477.85

About This Item

Empirical Formula (Hill Notation):
C21D5H27O5
Molecular Weight:
369.51
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

$477.85

List Price$503.00
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request more information

isotopic purity

≥98 atom % D

Quality Level

assay

≥98% (CP)

form

liquid

concentration

100 μg/mL in methanol

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C

SMILES string

OCC([C@@]1(O)CCC([C@]1(C)C2)C3CC[C@]4([H])C([2H])([2H])[C@]([2H])(O)C([2H])([2H])C[C@]4(C)C3C2=O)=O

InChI

1S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,18,22-23,26H,3-11H2,1-2H3/t12-,13-,14?,15?,18?,19+,20+,21+/m1/s1/i5D2,9D2,13D

InChI key

SYGWGHVTLUBCEM-GDDQKZIGSA-N

Related Categories

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service