Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809179

Sigma-Aldrich

N-Ethyl-N-isopropylpropan-2-aminium 3-(3-Chloro-2,5,6-trifluoropyridin-4-yl)-4-oxo-1,5-dioxaspiro[5.5]-undec-2-en-2-olate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30ClF3N2O4
CAS Number:
Molecular Weight:
478.93
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

powder

mp

118-123 °C

storage temp.

2-8°C

SMILES string

FC1=C(F)N=C(F)C(Cl)=C1C2=C([O-])OC3(CCCCC3)OC2=O.CC[NH+](C(C)C)C(C)C

InChI

1S/C14H11ClF3NO4.C8H19N/c15-8-6(9(16)11(18)19-10(8)17)7-12(20)22-14(23-13(7)21)4-2-1-3-5-14;1-6-9(7(2)3)8(4)5/h20H,1-5H2;7-8H,6H2,1-5H3

InChI key

PCXHULBKBXNUIT-UHFFFAOYSA-N

General description

N-Ethyl-N-isopropylpropan-2-aminium 3-(3-chloro-2,5,6-trifluoropyridin-4-yl)-4-oxo-1,5-dioxaspiro[5.5]-undec-2-en-2-olate is a fluoroarylated-meldrum′s acid adduct. It can be prepared by reacting cyclohexyl-Meldrum′s acid with 3-chloro-2,4,5,6- tetrafluoropyridine.[1]

Application

This compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameera M Senaweera et al.
The Journal of organic chemistry, 79(21), 10466-10476 (2014-10-02)
This work describes the facile and mono-selective per- and polyfluoroarylation of Meldrum's acid to generate a versatile synthon for highly fluorinated α-phenyl acetic acid derivatives, which provide straightforward access to fluorinated building blocks. The reaction takes place quickly, and most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service