809128
(R)-Ph-Quinox
Synonym(s):
2-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl]quinoline
About This Item
assay
≥95%
form
powder
reaction suitability
reagent type: catalyst
SMILES string
C1(C2=CC=C(C=CC=C3)C3=N2)=N[C@H](C4=CC=CC=C4)CO1
InChI
1S/C18H14N2O/c1-2-6-13(7-3-1)17-12-21-18(20-17)16-11-10-14-8-4-5-9-15(14)19-16/h1-11,17H,12H2/t17-/m0/s1
InChI key
OIFADUQNTPUCKK-KRWDZBQOSA-N
General description
Application
related product
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service