808830
Gold nanoparticles
15 nm diameter, biotin terminated, PEG 5000, OD 50, dispersion in H2O
Synonym(s):
Biotin Gold
About This Item
Recommended Products
material
PEG 5000
Quality Level
form
dispersion in H2O
nanoparticles
composition
, 8.19E13 particles/mL
packaging
pkg of 0.5 mL
storage condition
do not freeze
concentration
2.81 mg/mL in H2O
OD
50
surface area
20.6 m2/g
diameter
15 nm
pH
5-7
density
1.00 g/cm3
peak SPR wavelength
520 nm
functional group
biotin
storage temp.
2-8°C
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service