Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808733

Sigma-Aldrich

Tantalum diselenide

Crystal, 99.995%

Synonym(s):

Tantalum selenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
TaSe2
CAS Number:
Molecular Weight:
338.87
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.995%

form

crystals

L × W × H

0.5 cm × 0.5 cm × 0.1-0.15 mm, typical size, some variation in dimensions may occur with different batches

SMILES string

[Se]=[Ta]=[Se]

InChI

1S/2Se.Ta

InChI key

IYJABVNLJXJBTP-UHFFFAOYSA-N

General description

The hexagonal layered crystal structure of Tantalum diselenide constitutes of Se-Ta-Se layers in various stacking arrangements. The layered structure allows lamination or exfoliation of the layers.[1][2]

Application

The two dimensional layered inorganic nanomaterials have drawn significant attention recently owing to their thickness dependent physical and chemical properties. They find applications in photocatalysis, photovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc. Transition metal dischalcogenides represent an important class of inorganic 2D materials that have the aforementioned applications.[3][4][5]

Preparation Note

Store under Nitrogen atmosphere.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phonon and thermal properties of exfoliated TaSe2 thin films.
Yan Z, et al.
Journal of Applied Physics, 114(20), 204301-204301 (2013)
Temperature-dependent Fermi surface of 2 H-TaSe 2 driven by competing density wave order fluctuations.
Inosov DS, et al.
Physical Review. B, Condensed Matter and Materials Physics, 79(12) (2009)
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar JL, et al.
The Journal of Physical Chemistry C, 118, 3847-3863 (2014)

Articles

Advances in scalable synthesis and processing of two-dimensional materials

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service