Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808466

Sigma-Aldrich

4-arm Poly(ethylene glycol) norbornene terminated

average Mn 20,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C8H9O2(C2H4O)nCH2]4C
CAS Number:
UNSPSC Code:
12162002

form

powder

mol wt

average Mn 20,000

mp

49-54 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Multifunctional PEG for crosslinking, may be used to form hydrogels for biomaterial applications.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alex A Aimetti et al.
Biomaterials, 30(30), 6048-6054 (2009-08-14)
Degradable hydrogels have been extensively used in biomedical applications such as drug delivery, and recent interest has grown in hydrogels that degrade in recognition of a cellular response. This contribution describes a poly(ethylene glycol) (PEG) hydrogel platform with human neutrophil
Kelly C Santos Roballo et al.
Biomaterials, 209, 1-9 (2019-04-26)
Segmental injuries to peripheral nerves (PNs) too often result in lifelong disability or pain syndromes due to a lack of restorative treatment options. For injuries beyond a critical size, a bridging device must be inserted to direct regeneration. PN allografts

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service