Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808431

Sigma-Aldrich

Methylammonium iodide

0.42 M in 2-propanol

Synonym(s):

Methanamine hydriodide, Methylamine hydroiodide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

liquid

concentration

0.42 M in 2-propanol
0.42 M±0.04 M

Application

Methanamine hydriodide is an important precursor for the preparation of perovskite photoactive layers for solar energy conversion.[1][2][3]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F

flash_point_c

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David T Moore et al.
Journal of the American Chemical Society, 137(6), 2350-2358 (2015-01-28)
Methylammonium lead halide perovskite solar cells continue to excite the research community due to their rapidly increasing performance which, in large part, is due to improvements in film morphology. The next step in this progression is control of the crystal
Antonio Abate et al.
Nano letters, 14(6), 3247-3254 (2014-05-03)
Organic-inorganic halide perovskites, such as CH3NH3PbX3 (X = I(-), Br(-), Cl(-)), are attracting growing interest to prepare low-cost solar cells that are capable of converting sunlight to electricity at the highest efficiencies. Despite negligible effort on enhancing materials' purity or
Samuel D Stranks et al.
Nature nanotechnology, 10(5), 391-402 (2015-05-08)
Metal-halide perovskites are crystalline materials originally developed out of scientific curiosity. Unexpectedly, solar cells incorporating these perovskites are rapidly emerging as serious contenders to rival the leading photovoltaic technologies. Power conversion efficiencies have jumped from 3% to over 20% in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service