Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808415

Sigma-Aldrich

Magnesium borohydride

α phase

Synonym(s):

α-Magnesium borohydride, Magnesium tetrahydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(BH4)2
CAS Number:
Molecular Weight:
53.99
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

assay

≥95%

form

powder

reaction suitability

reagent type: reductant

SMILES string

[BH4-].[BH4-].[Mg++]

InChI

1S/2BH4.Mg/h2*1H4;/q2*-1;+2

InChI key

BAJRVYFABMRJEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium borohydride is one of the intensively investigated complex metal hydride. Due to it′s high gravimetric energy it finds applications as hydrogen storage material.[1][2]

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 2

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen cycling in γ-Mg(BH4)2 with cobalt-based additives.
Zavorotynska O, et al.
Journal of Material Chemistry A, 3, 6592-6592 (2015)
Nishiyama N.
Journal of the Society of Inorganic Materials, Japan, 21, 405-405 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service