Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808148

Sigma-Aldrich

TcBoc-Leu-OH

≥95%

Synonym(s):

N-(2,2,2-Trichloro-tert-butoxy)carbonyl-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18Cl3NO4
CAS Number:
Molecular Weight:
334.62
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

form

powder

reaction suitability

reaction type: C-H Activation
reagent type: ligand
reaction type: Peptide Synthesis

mp

108-113 °C

application(s)

peptide synthesis

functional group

amine
carboxylic acid

InChI

1S/C11H18Cl3NO4/c1-6(2)5-7(8(16)17)15-9(18)19-10(3,4)11(12,13)14/h6-7H,5H2,1-4H3,(H,15,18)(H,16,17)/t7-/m0/s1

InChI key

LMPYDSRXIGDZAB-ZETCQYMHSA-N

Application

Amino acid as a chiral ligand for enantioselective activation of C-H bond.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd(II)-catalyzed enantioselective activation of C(sp2)-H and C(sp3)-H bonds using monoprotected amino acids as chiral ligands.
Bing-Feng Shi et al.
Angewandte Chemie (International ed. in English), 47(26), 4882-4886 (2008-05-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service