Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

808016

Sigma-Aldrich

(R,S,S)-Elenor Amination Reagent

Synonym(s):

3H-3a,6-Methano-2,1-benzisothiazole-1(4H)-carboxamide, tetrahydro-N-hydroxy-8,8-dimethyl-, 2,2-dioxide, (3aR,6S,7aS)-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O4S
CAS Number:
Molecular Weight:
274.34
UNSPSC Code:
12352000
PubChem Substance ID:

form

powder

functional group

amine
sulfonamide

storage temp.

2-8°C

SMILES string

O=C(N1S(C[C@@]2(C3(C)C)CC[C@@]3([H])C[C@]12[H])(=O)=O)NO

InChI

1S/C11H18N2O4S/c1-10(2)7-3-4-11(10)6-18(16,17)13(8(11)5-7)9(14)12-15/h7-8,15H,3-6H2,1-2H3,(H,12,14)/t7-,8-,11-/m0/s1

InChI key

FYSORZDAOUCPGD-LAEOZQHASA-N

Application

The following hydroxyamide is a chiral electrophilic nitroso reagent for the transfer of nitrogen to enol ethers. This methology was reported by the Read de Alaniz Group, is a mild approach that can be performed at room temperature and in the presence of air.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service