Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

807818

Sigma-Aldrich

PCy3-Pd-G2 GT capsule

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43ClNPPd
Molecular Weight:
590.52
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

capsule

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

InChI

1S/C18H33P.C12H10N.ClH.Pd/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;;/h16-18H,1-15H2;1-6,8-9H,13H2;1H;/q;;;+1/p-1

InChI key

CYJRABDADOSMKA-UHFFFAOYSA-M

Application

Convenient dosable form of PCy3-Pd-G2.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service