Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807753

Sigma-Aldrich

Tungsten oxide nanoparticle ink

2.5 wt. %, viscosity 8 cP 

Synonym(s):

Avantama P-10-Jet, Nanograde P-10-Jet, W03 nanoparticle ink, WO3 ink, WO3 nanoparticle dispersion, WO3 nanoparticle suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

description

Solid content: 2.5 wt% crystalline WO3 in 2-propanol and propylene glycol
Viscosity: 6.1 ~ 10.1cP

form

dispersion

concentration

2.5 wt. %

work function

-5.3--5.7 eV

particle size

11-21 nm

viscosity

8 cP

Application

The WO3-x nanoparticle ink is for inkjet printing for the use as hole transport layer in printed electronics. The WO3-x nanoparticle ink is universally applicable in normal and inverted architecture solar cells.

Preparation Note

  • Storage: In dark at room temperature, Keep in plastic container (glass container can influence dispersion stability).
  • Prior to application: Shake, ultrasonicate with sonic horn.
  • Post-treatment: Annealing of deposited WO3 films at >120°C.
Upon need of ink dilution for viscosity/concentration tunability, 2-propanol can be used as a diluting solvent.

Legal Information

Product of Avantama Ltd.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

69.8 °F

flash_point_c

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Fill Factor Polymer Solar Cells Incorporating a Low Temperature Solution Processed WO3 Hole Extraction Layer
Christoph J, and Brabec et al.
Advanced Energy Materials, 2, 1433?1438-1433?1438 (2012)
Inverted structure organic photovoltaic devices employing a low temperature solution processed WO3 anode buffer layer
Christoph J, and Brabec et al.
Organic Electronics, 13(11), 2479?2484-2479?2484 (2012)

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service