Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807621

Sigma-Aldrich

Zinc oxide nanoparticle ink

2.5 wt. %, viscosity 30 cP, work function -4.3eV

Synonym(s):

Avantama N-10-Screen, Nanograde N-10-Screen, ZnO ink, ZnO nanoparticle ink, zinc oxide dispersion, zinc oxide suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion

concentration

2.5 wt. % (crystalline ZnO in α-terpineol)

work function

-4.1--4.5 eV

color

translucent brownish

particle size

8-16 nm

viscosity

26-36 cP

Looking for similar products? Visit Product Comparison Guide

Application

ZnO nanoparticle ink is for screen printing for the use as electron transport layer in printed electronics and it is universally applicable in normal and inverted architecture.

Preparation Note

  • Storage: In dark at room temperature.
  • Prior to application: Shake, ultrasonicate with sonic horn and (optionally) filter through 0.45 μm PTFE filter.
  • Post-treatment: Annealing of deposited ZnO films at >120°C.

Legal Information

Product of Avantama Ltd.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

197.1 °F

flash_point_c

91.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service