Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

807362

Sigma-Aldrich

Silver nanospheres

80 nm avg. part. size, 1 mg/mL in water, lipoic acid functionalized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.23

form

nanospheres

concentration

1 mg/mL in water

avg. part. size

80 nm

λmax

470 nm

functional group

lipoic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle′s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale “antennas” that can amplify the strength of the local electromagnetic field.

Preparation Note

Shake well before use.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service