806978
Silver nanospheres
10 nm avg. part. size, 1 mg/mL (aqueous sodium citrate), citrate functionalized
About This Item
form
nanospheres
concentration
1 mg/mL (aqueous sodium citrate)
avg. part. size
10 nm
λmax
390 nm
functional group
citrate
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Preparation Note
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service