806846
Silver
Nanowires, diam. × L 25 nm (±3 nm) × 15 μm (±2 μm), 5 mg/mL (in IPA)
About This Item
description
Flash point 53.6 °F
Quality Level
form
suspension
concentration
5 mg/mL (in IPA)
diam. × L
25 nm (±3 nm) × 15 μm (±2 μm)
color
gray
density
0.796 g/mL
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
53.6 °F - closed cup
flash_point_c
12.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service