Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

806676

Sigma-Aldrich

N-Methoxycarbonyl-Leu-OH

Synonym(s):

N-Methoxycarbonyl-Leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO4
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

form

solid

application(s)

peptide synthesis

SMILES string

O=C(O)[C@H](CC(C)C)NC(OC)=O

InChI

1S/C8H15NO4/c1-5(2)4-6(7(10)11)9-8(12)13-3/h5-6H,4H2,1-3H3,(H,9,12)(H,10,11)/t6-/m0/s1

InChI key

NPRGOLCTGVFFGS-LURJTMIESA-N

General description

N-Methoxycarbonyl-Leu-OH or N-methoxycarbonyl-leucine is a modified amino acid that can be employed as a building block in peptide synthesis.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service