806323
Hyperbranched G4-PEG20k-OH
Synonym(s):
bis-MPA hyperbranched PEG20k-OH dendrimer, pseudo generation 4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C1060H2062O546
Molecular Weight:
23545.39
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
powder
mp
56-61 °C
storage temp.
−20°C
General description
- End Group Functionality: Hydroxyl surface group
- Generation: Pseudo generation 4
- No. Surface Groups: 32
- Calculated Mol. Wt.: 23,545 g/mol
- Core: Polyethylene Glycol 20,000
- Branching units: 2,2-bis(methylol)propionic acid (bis-MPA)
Application
Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Multifunctional Poly(ethylene glycol): synthesis, characterization, and potential applications of dendritic-linear-dendritic block copolymer hybrids.
Andren O, et al.
Macromolecules, 46(10), 3726-3736 (2013)
Articles
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service