Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

806277

Sigma-Aldrich

Hyperbranched G5-PEG10k-OH

Synonym(s):

bis-MPA hyperbranched PEG10k-OH dendrimer, pseudo generation 5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C764H1406O414
Molecular Weight:
17217.09
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mp

48-53 °C

storage temp.

−20°C

General description

  • End Group Functionality: Hydroxyl surface group
  • Generation: Pseudo generation 5
  • No. Surface Groups: 64
  • Calculated Mol. Wt.: 17217 g/mol
  • Core: Polyethylene Glycol 10,000
  • Branching units: 2,2-bis(methylol)propionic acid (bis-MPA)

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional Poly(ethylene glycol): synthesis, characterization and potential applications of dendritic-linear-dendritic block copolymer hybrids.
Andren O, et al.
Macromolecules, 46, 3726-3736 (2013)

Articles

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service