Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

806161

Sigma-Aldrich

bis-MPA-Acetylene dendrimer

trimethylol propane core, generation 5

Synonym(s):

G5-TMP-Acetylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C1143H1334O570
Molecular Weight:
24192.48
UNSPSC Code:
12162002
NACRES:
NA.23

assay

95%

form

solid

storage temp.

−20°C

General description

  • End Group Functionality: Acetylene surface group
  • Generation: 5
  • No. Surface Groups: 96
  • Calculated Mol. Wt.: 5357.4 g/mol
  • Other Notes: 24192.7 g/mol

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service