Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

805653

Sigma-Aldrich

RuPhos-Pd-G3 GT capsule

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H56NO5PPdS
Molecular Weight:
836.37
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

capsule

feature

generation 3

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

CC(C)OC1=CC=CC(OC(C)C)=C1C2=CC=CC=C2[P+](C3CCCCC3)([Pd-2]4(OS(C)(=O)=O)C(C=CC=C5)=C5C6=C([NH2+]4)C=CC=C6)C7CCCCC7

InChI

1S/C30H43O2P.C12H10N.CH4O3S.Pd/c1-22(2)31-27-19-13-20-28(32-23(3)4)30(27)26-18-11-12-21-29(26)33(24-14-7-5-8-15-24)25-16-9-6-10-17-25;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h11-13,18-25H,5-10,14-17H2,1-4H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

AXZLIMCJMPNFFU-UHFFFAOYSA-M

Application

Convenient dosable form of RuPhos-Pd-G3

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The advent of the Buchwald portfolio of ligands and complexes for cross-coupling reactions in the past decade has greatly advanced the ease of performing this transformation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service