805084
Reduced graphene oxide
octadecylamine functionalized
Synonym(s):
rGO-ODA, rGO
About This Item
Recommended Products
form
powder (black)
composition
Carbon, >78 wt. %
Nitrogen, >3 wt. %
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
conductivity
6.36 S/m (pressed pellets)
solubility
chloroform: soluble
dichloromethane: soluble
greener alternative category
, Enabling
SMILES string
CNCCCCCCCCCCCCCCCCC.[rGO]
General description
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.
Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.
Advances in scalable synthesis and processing of two-dimensional materials
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service