All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H10BF2NO4
Molecular Weight:
269.01
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
form
powder
mp
228.4 °C
storage temp.
2-8°C
SMILES string
O=C(CN(C)C1)OB(C2=C(F)C(F)=CC=C2)OC1=O
InChI
1S/C11H10BF2NO4/c1-15-5-9(16)18-12(19-10(17)6-15)7-3-2-4-8(13)11(7)14/h2-4H,5-6H2,1H3
InChI key
UPYJOVJUDRUPPY-UHFFFAOYSA-N
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service