Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

804932

Sigma-Aldrich

2-(1-Phenyl-1H-1,2,3-triazol-4-yl)-boronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13BN4O4
Molecular Weight:
300.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

195 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2cn(nn2)-c3ccccc3

InChI

1S/C13H13BN4O4/c1-17-8-12(19)21-14(22-13(20)9-17)11-7-18(16-15-11)10-5-3-2-4-6-10/h2-7H,8-9H2,1H3

InChI key

HTDHAOUYVRWZCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service