Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

804908

Sigma-Aldrich

DCJTB

Synonym(s):

4-(dicyanomethylene)-2-tert-butyl-6-(1,1,7,7-tetramethyljulolidin-4-yl-vinyl)-4H-pyran, 4-(Dicyanomethylene)-2-tert-butyl-6-(1,1,7,7-tetramethyljulolidin-4-yl-vinyl)-4H-pyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H35N3O
CAS Number:
Molecular Weight:
453.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

99% (HPLC)

form

powder

mp

298-303 °C (exp.)

orbital energy

HOMO 5.3 eV 
LUMO 3.1 eV 

OLED device performance

Glass/CCL(DCJTB 0.2 wt%)/ITO/PEDOT:PSS/ [Ir(dfppz)2(dtb-bpy)]+ (PF6 )/Ag (100 nm)

  • Max. EQE: 14.1 %
  • Life Time: 11 min.

ITO/MoO3/m-MTDATA/mMTDATA:BPhen:DCJTB(2 vol%)/BPhen/LiF/Al
  • Color: red
  • Turn-On Voltage: 2.3 V

SMILES string

CC(CC1)(C)C2=C(N1CCC3(C)C)C3=CC(/C=C/C4=CC(C=C(C(C)(C)C)O4)=C(C#N)C#N)=C2

InChI

1S/C30H35N3O/c1-28(2,3)26-17-21(22(18-31)19-32)16-23(34-26)9-8-20-14-24-27-25(15-20)30(6,7)11-13-33(27)12-10-29(24,4)5/h8-9,14-17H,10-13H2,1-7H3/b9-8+

InChI key

HXWWMGJBPGRWRS-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DCJTB belongs to a class of laser dyes useful for solar-concentrator applications.[1] Pure DCJTB can be prepared from unsymmetrical 4-(dicyanomethylene)-2-(t-butyl)-6-methyl-4H-pyran directly. [2] These red dopants have high photoluminescent (PL) quantum yields. Its photoluminescent properties have been reported in a study. [3]

Application

Red dopant for white organic light emitting diodes (OLEDs). 1[4] DCJTB may be used as an emitting layer to enhance the performance of OLEDs. [3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trap effect of an ultrathin DCJTB layer in organic light-emitting diodes
Wang YM, et al.
Materials Chemistry and Physics null
White top-emitting organic light-emitting diodes using one-emissive layer of the DCJTB doped DPVBi layer
Kim MS, et al.
Thin Solid Films, 3590-3594 null
Red fluorescent organic light-emitting diodes using modified pyran-containing DCJTB derivatives.
Lee KH, et al.
Bull. Korean Chem. Soc. null
Trap effect of an ultrathin DCJTB layer in organic light-emitting diodes
Wang YM, et al.
Materials Chemistry and Physics null
A New Family of Red Dopants Based on Chromene-Containing Compounds for Organic Electroluminescent Devices.
Zhang et al.
Journal of Material Chemistry C, 13(5), 1565-1569 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service