Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

804894

Sigma-Aldrich

H101

Synonym(s):

2,5-bis(4,4’-bis(methoxyphenyl)aminophen-4’’-yl)-3,4-ethylenedioxythiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H40N2O6S
CAS Number:
Molecular Weight:
748.88
MDL number:
UNSPSC Code:
12352103

assay

>98%

form

powder

mp

159-164 °C

SMILES string

COC(C=C1)=CC=C1N(C2=CC=C(OC)C=C2)C(C=C3)=CC=C3C4=C5C(OCCO5)=C(C6=CC=C(N(C7=CC=C(OC)C=C7)C8=CC=C(OC)C=C8)C=C6)S4

Application

A novel electron-rich 3,4-ethylenedioxythiophene based hole transporting material for perovskite solar cells with the PCE of 13.8% under AM 1.5G solar simulation.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple 3,4-Ethylenedioxythiophene Based Hole-Transporting Material for Perovskite Solar Cells
Li H, et al.
Angewandte Chemie (International Edition in English), 53(16), 4085?4088-4085?4088 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service