Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

804886

Sigma-Aldrich

B3PyPB

Synonym(s):

1,3-Bis(3,5-dipyrid-3-ylphenyl)benzene, 1,3-Bis[3,5-di(pyridin-3-yl)phenyl]benzene, 3,3′,3′′,3′′′-[1,1′:3′,1′′-terphenyl]-3,3′′,5,5′′-tetrayltetrakispyridine, 3,5,3′′,5′′-Tetra-3-pyridyl-1,1′:3,1′′-terphenyl, BmPyPhB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H26N4
CAS Number:
Molecular Weight:
538.64
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥99% (HPLC)

form

powder

mp

264-269 °C

SMILES string

C1(C2=CC(C3=CC=CN=C3)=CC(C4=CN=CC=C4)=C2)=CC=CC(C5=CC(C6=CC=CN=C6)=CC(C7=CN=CC=C7)=C5)=C1

InChI

1S/C38H26N4/c1-6-27(33-17-35(29-8-2-12-39-23-29)21-36(18-33)30-9-3-13-40-24-30)16-28(7-1)34-19-37(31-10-4-14-41-25-31)22-38(20-34)32-11-5-15-42-26-32/h1-26H

InChI key

WCXKTQVEKDHQIY-UHFFFAOYSA-N

Application

Wide energy gap (~ 4.05 eV) electron transport materials containing 3, 5-dipyridylphenyl moieties for high efficiency PHOLED.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and Stable White Organic Light-Emitting Diodes Employing a Single Emitter.
Li G, et al.
Advanced Materials, 26(18), 2931-2936 (2014)
Simplified organic light-emitting devices utilizing ultrathin electron transport layers and new insights on their roles.
Zhang Y, et al.
ACS Applied Materials & Interfaces, 6(3), 1697-1701 (2014)
Wide-Energy-Gap Electron-Transport Materials Containing 3,5-Dipyridylphenyl Moieties for an Ultra High Efficiency Blue Organic Light-Emitting Device.
Hisahiro S, et al.
Chemistry of Materials, 20(19), 5951-5953 (2008)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service